Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284377.1 | NP_001271306.1 | 3 | 206 | ||
NM_001284378.1 | NP_001271307.1 | 3 | 206 | |||
NM_001284379.1 | NP_001271308.1 | 2 | 184 | |||
NM_001321844.1 | NP_001308773.1 | 3 | 200 | |||
NM_001321848.1 | NP_001308777.1 | 2 | 184 | |||
NM_017828.4 | NP_060298.2 | 3 | 206 | |||
NR_104312.1 | 3 | 206 | ||||
NR_104313.1 | 3 | 206 | ||||
XM_005254509.3 | XP_005254566.1 | 3 | 232 | |||
XM_011521739.1 | XP_011520041.1 | 3 | 241 | |||
XM_011521741.1 | XP_011520043.1 | 3 | 241 | |||
GenBank mRNA | AK000459.1 | - | 3 | 143 | ||
AK057145.1 | - | 2 | 359 | |||
AK225716.1 | - | 3 | 167 | |||
AK293708.1 | - | 3 | 142 | |||
AK314740.1 | - | 3 | 139 | |||
AL136872.1 | - | 3 | 126 | |||
AY542160.1 | - | 2 | 112 | |||
BC000837.2 | - | 2 | 109 | |||
BC033744.1 | - | 2 | 378 | |||
BC068998.1 | - | 3 | 128 | 6586373 | ||
BC069195.1 | - | 3 | 134 | |||
BI829264.1 | - | 3 | 182 | |||
BI829484.1 | - | 3 | 206 | |||
BQ233598.1 | - | 169 | ||||
BU540913.1 | - | 3 | 148 | |||
CR533452.1 | - | 2 | 112 | |||
HY162014.1 | - | 2 | 185 |
Gene Symbol: GOLGA6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022406.1 | XP_016877895.1 | 2 | 642 |
Gene Symbol: LOC440292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522301.2 | XP_011520603.1 | 2 | 216 | ||
XM_017022799.1 | XP_016878288.1 | 2 | 636 |
Gene Symbol: LOC732265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022776.1 | XP_016878265.1 | 3 | 554 |