Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018303.5 | NP_060773.3 | 4 | 533 | ||
NR_073064.1 | 6 | 695 | ||||
XM_017011018.1 | XP_016866507.1 | 5 | 579 | |||
XM_017011019.1 | XP_016866508.1 | 5 | 741 | |||
XM_017011020.1 | XP_016866509.1 | 5 | 601 | |||
XM_017011021.1 | XP_016866510.1 | 4 | 837 | |||
XM_017011022.1 | XP_016866511.1 | 4 | 533 | |||
XM_017011023.1 | XP_016866512.1 | 4 | 533 | |||
XM_017011024.1 | XP_016866513.1 | 5 | 579 | |||
XM_017011025.1 | XP_016866514.1 | 4 | 533 | |||
XM_017011026.1 | XP_016866515.1 | 4 | 533 | |||
GenBank mRNA | AJ414403.1 | - | 4 | 457 | ||
AK056564.1 | - | 4 | 501 | |||
AK314628.1 | - | 4 | 533 | |||
AL833209.1 | - | 2 | 89 | |||
AL833213.1 | - | 4 | 479 | |||
BC016918.1 | - | 1 | 13 | |||
BC080542.1 | - | 6 | 665 | 5479496 | ||
DA228027.1 | - | 4 | 533 |