Search
Search

FAM219B| Gene Symbol: FAM219B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321920.1 | NP_001308849.1 | 3 | 423 | ||
| NM_001321921.1 | NP_001308850.1 | 3 | 423 | |||
| NM_001321922.1 | NP_001308851.1 | 3 | 423 | |||
| NM_001321923.1 | NP_001308852.1 | 3 | 423 | |||
| NM_020447.4 | NP_065180.1 | 3 | 423 | |||
| NR_135841.1 | 2 | 335 | ||||
| NR_135842.1 | 2 | 335 | ||||
| NR_135843.1 | 2 | 335 | ||||
| NR_135844.1 | 2 | 335 | ||||
| NR_135845.1 | 3 | 423 | ||||
| NR_135846.1 | 3 | 423 | ||||
| NR_135847.1 | 2 | 335 | ||||
| NR_135848.1 | 2 | 335 | ||||
| NR_135849.1 | 3 | 423 | ||||
| NR_135850.1 | 2 | 335 | ||||
| NR_135851.1 | 3 | 423 | ||||
| NR_135852.1 | 2 | 335 | ||||
| NR_135853.1 | 3 | 423 | ||||
| NR_135854.1 | 2 | 335 | ||||
| XM_017022433.1 | XP_016877922.1 | 3 | 745 | |||
| GenBank mRNA | AK000005.1 | - | 2 | 1371 | ||
| AK001572.1 | - | 2 | 324 | |||
| AK291020.1 | - | 3 | 410 | |||
| AK296403.1 | - | 3 | 410 | |||
| AK308954.1 | - | 2 | 319 | |||
| BC015637.2 | - | 3 | 367 | 4636850 | ||
| DB153105.1 | - | 3 | 423 | |||