Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021073.2 | NP_066551.1 | 1 | 855 | ||
XM_005249304.3 | XP_005249361.1 | 1 | 903 | |||
XM_011514816.2 | XP_011513118.1 | 1 | 151 | |||
XM_011514817.2 | XP_011513119.1 | 1 | 903 | |||
GenBank mRNA | AK291559.1 | - | 1 | 868 | ||
AK303576.1 | - | 1 | 157 | |||
BC027958.1 | - | 1 | 855 | 5209637 | ||
KC485577.1 | - | 1 | 143 | |||
M60314.1 | - | 1 | 841 |