Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABR SIRNA Assays ›
          Gene Symbol:
          ABR
          Gene Name:
          active BCR-related
          Gene Aliases:
          MDB
          Chromosome Location:
          Chr.17: 1003518 - 1229713 on Build GRCh38
          UniGene ID:
          Hs.159306
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 147814

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001092.4 NP_001083.2 9 949
          NM_001159746.2 NP_001153218.1 10 1490
          NM_001282149.1 NP_001269078.1 5 661
          NM_001322840.1 NP_001309769.1 10 1193
          NM_001322841.1 NP_001309770.1 10 1855
          NM_021962.4 NP_068781.2 10 1275
          XM_011523810.2 XP_011522112.1 10 1078
          XM_011523811.2 XP_011522113.2 10 1888
          XM_011523812.2 XP_011522114.1 9 2425
          XM_011523813.2 XP_011522115.1 9 949
          XM_011523814.2 XP_011522116.1 10 1490
          XM_011523815.2 XP_011522117.1 10 1173
          XM_017024539.1 XP_016880028.1 10 1885
          XM_017024540.1 XP_016880029.1 9 1787
          XM_017024541.1 XP_016880030.1 10 1888
          XM_017024542.1 XP_016880031.1 5 646
          XM_017024543.1 XP_016880032.1 5 656
          GenBank mRNA AK124547.1 - 10 1490
          AK126882.1 - 10 1077
          AK295191.1 - 9 867
          AK299909.1 - 5 516
          AK302682.1 - 10 1016
          AK307827.1 - 9 1354
          DA327723.1 - 3 258
          U01147.1 - 10 1138

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cellular extravasation
          small GTPase mediated signal transduction
          brain development
          actin cytoskeleton organization
          response to lipopolysaccharide
          regulation of Rho protein signal transduction
          inner ear morphogenesis
          positive regulation of apoptotic process
          regulation of vascular permeability
          negative regulation of neutrophil degranulation
          positive regulation of GTPase activity
          negative regulation of inflammatory response
          positive regulation of phagocytosis
          neuromuscular process controlling balance
          regulation of small GTPase mediated signal transduction
          negative regulation of blood vessel remodeling
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          GTPase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0