Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024649.4 | NP_078925.3 | 11 | 1082 | ||
GenBank mRNA | AF503941.1 | - | 11 | 1082 | ||
AK095638.1 | - | 10 | 1089 | |||
AK294962.1 | - | 3 | 316 | |||
AK296311.1 | - | 7 | 797 | |||
BC047642.1 | - | 10 | 1041 | 4824649 | ||
BC109064.2 | - | 11 | 1075 | 40033833 | ||
BX647612.1 | - | 11 | 1111 |