Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146343.1 | NP_001139815.1 | 4 | 2102 | ||
NM_001146345.1 | NP_001139817.1 | 5 | 2319 | |||
NM_025078.4 | NP_079354.2 | 6 | 2373 | |||
XM_005266770.2 | XP_005266827.1 | 5 | 2168 | |||
XM_011526188.1 | XP_011524490.1 | 4 | 1984 | |||
XM_017025997.1 | XP_016881486.1 | 7 | 2460 | |||
XM_017025998.1 | XP_016881487.1 | 6 | 2406 | |||
XM_017026001.1 | XP_016881490.1 | 5 | 2390 | |||
XM_017026003.1 | XP_016881492.1 | 6 | 2243 | |||
XM_017026004.1 | XP_016881493.1 | 5 | 2189 | |||
XM_017026005.1 | XP_016881494.1 | 5 | 2108 | |||
GenBank mRNA | AK123870.1 | - | 6 | 2246 | ||
AK126188.1 | - | 6 | 2764 | |||
BC030140.2 | - | 6 | 2339 | 4761671 |