Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BBS4 SIRNA Assays ›
          Gene Symbol:
          BBS4
          Gene Name:
          Bardet-Biedl syndrome 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 72686179 - 72738476 on Build GRCh38
          UniGene ID:
          Hs.208681
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 147959

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BBS4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252678.1 NP_001239607.1 5 360
          NM_001320665.1 NP_001307594.1 6 412
          NM_033028.4 NP_149017.2 6 412
          NR_045565.1 7 470
          NR_045566.1 6 725
          XM_011521848.1 XP_011520150.1 6 382
          XM_011521849.1 XP_011520151.1 5 275
          XM_011521851.1 XP_011520153.1 6 382
          XM_017022450.1 XP_016877939.1 6 693
          XM_017022451.1 XP_016877940.1 5 353
          XM_017022452.1 XP_016877941.1 5 583
          XM_017022453.1 XP_016877942.1 5 265
          XM_017022454.1 XP_016877943.1 4 215
          GenBank mRNA AF359281.1 - 6 384
          AK075321.1 - 6 376
          AK298922.1 - 7 442
          AK302224.1 - 6 679
          AK303706.1 - 5 355
          AK314507.1 - 6 384
          AY457143.1 - 6 484
          BC008923.2 - 3 189
          BC027624.2 - 6 382 4824276
          BE727117.1 - 5 353
          BI562463.1 - 6 402
          BX647855.1 - 5 331
          DB269838.1 - 6 405
          DC320069.1 - 6 407
          DC372967.1 - 6 412
          KU177936.1 - 6 363

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          mitotic cytokinesis
          neuron migration
          neural tube closure
          retina homeostasis
          heart looping
          negative regulation of systemic arterial blood pressure
          spermatid development
          visual perception
          sensory perception of smell
          negative regulation of gene expression
          protein transport
          dendrite development
          regulation of lipid metabolic process
          ventricular system development
          striatum development
          hippocampus development
          cerebral cortex development
          adult behavior
          negative regulation of actin filament polymerization
          melanosome transport
          regulation of cytokinesis
          protein localization to organelle
          negative regulation of GTPase activity
          microtubule anchoring at centrosome
          nonmotile primary cilium assembly
          social behavior
          photoreceptor cell outer segment organization
          negative regulation of appetite by leptin-mediated signaling pathway
          positive regulation of multicellular organism growth
          cilium assembly
          fat cell differentiation
          photoreceptor cell maintenance
          positive regulation of cilium assembly
          retinal rod cell development
          intracellular transport
          brain morphogenesis
          sensory processing
          centrosome organization
          maintenance of protein location in nucleus
          regulation of stress fiber assembly
          cilium morphogenesis
          regulation of cilium beat frequency involved in ciliary motility
          face development
          fat pad development
          protein localization to cilium
          protein localization to centrosome
          regulation of nonmotile primary cilium assembly
          protein localization to photoreceptor outer segment
          RNA polymerase II repressing transcription factor binding
          microtubule motor activity
          protein binding
          dynactin binding
          alpha-tubulin binding
          beta-tubulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0