Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033117.3 | NP_149108.1 | 6 | 775 | ||
NR_027125.1 | 7 | 900 | ||||
NR_027126.1 | 7 | 869 | ||||
XM_006717319.1 | XP_006717382.1 | 6 | 731 | |||
GenBank mRNA | AK057676.1 | - | 6 | 751 | ||
AK301050.1 | - | 4 | 474 | |||
AK310008.1 | - | 7 | 857 | |||
AK310807.1 | - | 7 | 811 | |||
AL389986.1 | - | 6 | 703 | |||
BC008942.2 | - | 6 | 722 | 2822634 | ||
BC019319.1 | - | 6 | 722 | 2822634 |