Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BDNF SIRNA Assays ›
Gene Symbol:
BDNF
Gene Name:
brain derived neurotrophic factor
Gene Aliases:
ANON2, BULN2
Chromosome Location:
Chr.11: 27654893 - 27722058 on Build GRCh38
UniGene ID:
Hs.502182
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148175
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BDNF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143805.1
NP_001137277.1
2
420
NM_001143806.1
NP_001137278.1
2
635
NM_001143807.1
NP_001137279.1
2
521
NM_001143808.1
NP_001137280.1
2
357
NM_001143809.1
NP_001137281.1
2
367
NM_001143810.1
NP_001137282.1
3
650
NM_001143811.1
NP_001137283.1
4
767
NM_001143812.1
NP_001137284.1
2
509
NM_001143813.1
NP_001137285.1
2
591
NM_001143814.1
NP_001137286.1
3
716
NM_001143816.1
NP_001137288.1
2
1114
NM_001709.4
NP_001700.2
2
609
NM_170731.4
NP_733927.1
2
931
NM_170732.4
NP_733928.1
2
718
NM_170733.3
NP_733929.1
2
621
NM_170734.3
NP_733930.1
2
569
NM_170735.5
NP_733931.1
1
1348
XM_011520280.2
XP_011518582.1
2
1515
GenBank mRNA
AF400438.1
-
1
281
AK289763.1
-
2
417
AK289853.1
-
2
318
AY054392.2
-
2
622
AY054393.2
-
2
364
AY054394.2
-
2
579
AY054395.2
-
2
662
AY054396.2
-
2
420
AY054397.2
-
2
503
AY054398.2
-
2
550
AY054399.2
-
3
435
AY054400.2
-
2
536
AY054406.3
-
2
404
AY656701.1
-
1
263
BC029795.1
-
2
594
5193877
CX751420.1
-
2
488
EF674517.1
-
2
420
EF674518.1
-
2
635
EF674519.1
-
2
718
EF674520.1
-
2
521
EF674521.1
-
2
621
EF689009.1
-
1
1362
EF689010.1
-
2
1128
EF689011.1
-
2
357
EF689012.1
-
2
367
EF689013.1
-
2
509
EF689014.1
-
2
591
EF689015.1
-
2
609
EF689016.1
-
3
716
EF689017.1
-
3
468
EF689018.1
-
2
569
EF689019.1
-
3
650
EF689020.1
-
4
767
EF689021.1
-
2
931
KP229412.1
-
1
263
KU177943.1
-
1
263
KU177944.1
-
1
263
KU177945.1
-
1
263
M61176.1
-
1
825
X60201.1
-
2
559
X91251.1
-
2
547
Back To Top
More Information
Panther Classification:
Molecular Function -
neurotrophic factor
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell signaling
nervous system development
axon guidance
synapse assembly
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of brain-derived neurotrophic factor receptor signaling pathway
negative regulation of neuron apoptotic process
collateral sprouting
positive regulation of collateral sprouting
positive regulation of synapse assembly
neurotrophin TRKB receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0