Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DST SIRNA Assays ›
Gene Symbol:
DST
Gene Name:
dystonin
Gene Aliases:
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location:
Chr.6: 56457987 - 56954671 on Build GRCh38
UniGene ID:
Hs.604915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148254
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144769.2
NP_001138241.1
16
1900
NM_001144770.1
NP_001138242.1
14
1571
NM_001723.5
NP_001714.1
3
455
NM_015548.4
NP_056363.2
3
455
NM_183380.3
NP_899236.1
13
1845
XM_005249310.3
XP_005249367.1
17
1994
XM_005249315.3
XP_005249372.1
16
1941
XM_005249316.2
XP_005249373.1
9
2217
XM_005249318.2
XP_005249375.1
15
1655
XM_005249319.2
XP_005249376.1
14
1556
XM_005249320.4
XP_005249377.1
14
1612
XM_005249322.4
XP_005249379.1
14
1507
XM_005249323.3
XP_005249380.1
13
1840
XM_005249324.2
XP_005249381.1
3
452
XM_011514824.2
XP_011513126.1
16
1905
XM_011514825.2
XP_011513127.1
16
1901
XM_011514826.2
XP_011513128.1
13
1840
XM_017011205.1
XP_016866694.1
17
2004
XM_017011206.1
XP_016866695.1
17
2004
XM_017011207.1
XP_016866696.1
17
1941
XM_017011208.1
XP_016866697.1
17
2004
XM_017011209.1
XP_016866698.1
17
2004
XM_017011210.1
XP_016866699.1
17
2004
XM_017011211.1
XP_016866700.1
17
2004
XM_017011212.1
XP_016866701.1
17
2004
XM_017011213.1
XP_016866702.1
17
2004
XM_017011214.1
XP_016866703.1
17
2004
XM_017011215.1
XP_016866704.1
17
2004
XM_017011216.1
XP_016866705.1
17
2004
XM_017011217.1
XP_016866706.1
14
1585
XM_017011219.1
XP_016866708.1
17
2000
XM_017011220.1
XP_016866709.1
16
1943
XM_017011221.1
XP_016866710.1
17
2000
XM_017011222.1
XP_016866711.1
9
2159
XM_017011223.1
XP_016866712.1
17
2000
XM_017011224.1
XP_016866713.1
13
1840
XM_017011225.1
XP_016866714.1
17
2014
GenBank mRNA
AF400226.1
-
3
347
AF400227.1
-
3
347
AK294830.1
-
13
1453
AK295864.1
-
9
1827
AK299898.1
-
14
1571
BC065536.1
-
3
419
DA462108.1
-
3
455
DA786051.1
-
4
342
L11690.1
-
3
449
M69225.1
-
3
385
U31850.1
-
13
1845
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
cell adhesion
integrin-mediated signaling pathway
response to wounding
maintenance of cell polarity
intermediate filament cytoskeleton organization
cell motility
actin binding
integrin binding
calcium ion binding
protein binding
protein C-terminus binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0