Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM102A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035254.2 | NP_001030331.1 | 11 | 3780 | ||
NM_203305.2 | NP_976050.1 | 8 | 3262 | |||
GenBank mRNA | AK058004.1 | - | 4 | 1457 | ||
AK074108.1 | - | 5 | 4237 | |||
AL049365.1 | - | 1 | 2164 | |||
BC047949.1 | - | 8 | 3265 | |||
BC142952.1 | - | 6 | 3071 |