Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220484.1 | NP_001207413.1 | 3 | 641 | ||
NM_203309.2 | NP_976054.2 | 2 | 562 | |||
XM_006720143.2 | XP_006720206.1 | 2 | 599 | |||
XM_011536760.2 | XP_011535062.1 | 2 | 785 | |||
XM_011536761.2 | XP_011535063.1 | 2 | 444 | |||
XM_017021289.1 | XP_016876778.1 | 2 | 1123 | |||
GenBank mRNA | AK093515.1 | - | 2 | 555 | ||
AK302564.1 | - | 3 | 615 | |||
BC047590.1 | - | 2 | 562 | 4837558 | ||
GU727638.1 | - | 1 | 152 |