Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193644.1 | NP_001180573.1 | 6 | 770 | ||
NM_002857.3 | NP_002848.1 | 6 | 770 | |||
NR_036492.1 | 5 | 660 | ||||
NR_036493.1 | 5 | 684 | ||||
GenBank mRNA | AB018541.1 | - | 6 | 747 | ||
AB062286.1 | - | 5 | 630 | |||
AK300368.1 | - | 5 | 639 | |||
AK303099.1 | - | 5 | 679 | |||
BC000496.2 | - | 6 | 745 | 2820701 | ||
BT006879.1 | - | 6 | 743 | |||
CD557913.1 | - | 6 | 760 | |||
KU178290.1 | - | 6 | 743 | |||
KU178291.1 | - | 5 | 633 | |||
X75535.1 | - | 6 | 753 |