Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LDB3 SIRNA Assays ›
Gene Symbol:
LDB3
Gene Name:
LIM domain binding 3
Gene Aliases:
CMD1C, CMH24, CMPD3, CYPHER, LDB3Z1, LDB3Z4, LVNC3, MFM4, ORACLE, PDLIM6, ZASP
Chromosome Location:
Chr.10: 86664785 - 86736072 on Build GRCh38
UniGene ID:
Hs.657271
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148396
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LDB3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080114.1
NP_001073583.1
5, 6
582
NM_001080115.1
NP_001073584.1
4, 5
723
NM_001080116.1
NP_001073585.1
5, 6
687
NM_001171610.1
NP_001165081.1
6, 7
1049
NM_001171611.1
NP_001165082.1
5, 6
927
NM_007078.2
NP_009009.1
4, 5
723
XM_005269464.4
XP_005269521.1
5, 6
892
XM_005269466.4
XP_005269523.1
5, 6
891
XM_005269468.4
XP_005269525.1
5, 6
888
XM_011539184.2
XP_011537486.1
6, 7
1097
XM_011539185.2
XP_011537487.1
6, 7
2991
XM_011539186.2
XP_011537488.1
6, 7
1097
XM_011539187.2
XP_011537489.1
6, 7
1096
XM_011539188.2
XP_011537490.1
5, 6
893
XM_011539189.1
XP_011537491.1
6, 7
646
XM_011539190.1
XP_011537492.1
5, 6
582
XM_011539191.2
XP_011537493.1
6, 7
562
XM_011539193.1
XP_011537495.1
4, 5
419
XM_011539194.1
XP_011537496.1
4, 5
419
XM_011539195.2
XP_011537497.1
6, 7
1093
XM_017015606.1
XP_016871095.1
5, 6
2787
XM_017015607.1
XP_016871096.1
1, 2
1023
XM_017015608.1
XP_016871097.1
5, 6
2787
XM_017015609.1
XP_016871098.1
6, 7
646
GenBank mRNA
AB014513.1
-
4, 5
721
AF276807.1
-
4, 5
700
AF276808.1
-
6, 7
927
AF276809.1
-
5, 6
904
AJ133766.1
-
5, 6
638
AJ133767.1
-
5, 6
559
AK294696.1
-
5, 6
780
AK297871.1
-
5, 6
679
AK300692.1
-
6, 7
595
AK304760.1
-
6, 7
991
BC010929.2
-
5, 6
584
DA762127.1
-
1, 2
187
KF772970.1
-
4, 5
700
KU178543.1
-
5, 6
559
KU178544.1
-
4, 5
700
KU178545.1
-
3, 4
332
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
sarcomere organization
protein kinase C binding
protein binding
cytoskeletal protein binding
zinc ion binding
muscle alpha-actinin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0