Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004557.3 | NP_004548.3 | 4 | 769 | ||
NR_134949.1 | 4 | 769 | ||||
NR_134950.1 | 4 | 769 | ||||
GenBank mRNA | AK290785.1 | - | 4 | 766 | ||
BC063815.1 | - | 4 | 759 | |||
BC140782.1 | - | 4 | 724 | 9021650 | ||
BC144540.1 | - | 4 | 730 | |||
BC144541.1 | - | 4 | 724 | |||
U95299.1 | - | 4 | 720 |