Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SERPINB6 SIRNA Assays ›
          Gene Symbol:
          SERPINB6
          Gene Name:
          serpin family B member 6
          Gene Aliases:
          CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3
          Chromosome Location:
          Chr.6: 2948159 - 2972165 on Build GRCh38
          UniGene ID:
          Hs.519523
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 14877

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SERPINB6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195291.2 NP_001182220.2 5 1143
          NM_001271822.1 NP_001258751.1 4 568
          NM_001271823.1 NP_001258752.1 4 666
          NM_001271824.1 NP_001258753.1 4 1050
          NM_001271825.1 NP_001258754.1 4 652
          NM_001297699.1 NP_001284628.1 4 1293
          NM_001297700.1 NP_001284629.1 4 949
          NM_004568.5 NP_004559.4 4 1008
          XM_011514672.1 XP_011512974.1 6 946
          XM_011514673.1 XP_011512975.1 4 1100
          XM_011514674.2 XP_011512976.1 4 685
          XM_011514676.2 XP_011512978.1 4 495
          XM_017010940.1 XP_016866429.1 5 507
          XM_017010941.1 XP_016866430.1 3 2136
          GenBank mRNA AB209563.1 - 3 676
          AF116727.1 - 1 1229
          AK027149.1 - 3 1834
          AK057138.1 - 3 2367
          AK094983.1 - 5 1143
          AK098566.1 - 4 665
          AK122895.1 - 2 1271
          AK122961.1 - 3 1284
          AK128637.1 - 2 2697
          AK314578.1 - 4 727
          BC001394.2 - 4 590 3051381
          BC098564.1 - 4 446 30563105
          BG499326.1 - 4 525
          BI090002.1 - 4 650
          BI259189.1 - 4 470
          BM789272.1 - 4 631
          BP325314.1 - 4 445
          BP370484.1 - 4 501
          BQ681973.1 - 4 455
          BX375950.2 - 4 734
          BX538343.1 - 4 4720
          CX755337.1 - 4 571
          CX782163.1 - 4 518
          Z22658.1 - 4 439

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          sensory perception of sound
          negative regulation of endopeptidase activity
          cellular response to osmotic stress
          protease binding
          serine-type endopeptidase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0