siRNA Details
Gene Symbol: CRTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042574.2 | NP_001036039.1 | 15 | 2628 | ||
NM_022769.4 | NP_073606.3 | 15 | 2631 | |||
XM_005254968.2 | XP_005255025.1 | 15 | 2599 | |||
XM_011521906.2 | XP_011520208.1 | 14 | 2503 | |||
GenBank mRNA | AK025521.1 | - | 8 | 1795 | ||
AK074170.1 | - | 12 | 2099 | |||
AK090443.1 | - | 15 | 2588 | |||
AK225300.1 | - | 8 | 1795 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -



