Search
Search

GAL3ST4| Gene Symbol: GAL3ST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024637.4 | NP_078913.3 | 4 | 1604 | ||
| GenBank mRNA | AF289575.1 | - | 3 | 1399 | ||
| AF316113.1 | - | 4 | 1447 | |||
| AK022178.1 | - | 3 | 1243 | |||
| AK092367.1 | - | 3 | 2230 | |||
| AK225890.1 | - | 3 | 1380 | |||
| AK301591.1 | - | 2 | 1042 | |||
| AL833824.1 | - | 3 | 1249 | |||
| BC012976.2 | - | 4 | 1423 | 3636329 | ||