Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200049.2 | NP_001186978.2 | 7, 8 | 862 | ||
GenBank mRNA | AK098820.1 | - | 7, 8 | 837 | ||
BC146900.1 | - | 6, 7 | 645 | 9056828 | ||
BC146908.1 | - | 6, 7 | 645 | 9056836 |