Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RAPGEF1 SIRNA Assays ›
Gene Symbol:
RAPGEF1
Gene Name:
Rap guanine nucleotide exchange factor 1
Gene Aliases:
C3G, GRF2
Chromosome Location:
Chr.9: 131576770 - 131740068 on Build GRCh38
UniGene ID:
Hs.127897
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
149383
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RAPGEF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001304275.1
NP_001291204.1
24
3278
NM_005312.3
NP_005303.2
24
3237
NM_198679.1
NP_941372.1
24
3326
XM_005272186.4
XP_005272243.1
27
3912
XM_005272191.3
XP_005272248.1
26
3755
XM_006717067.3
XP_006717130.1
27
3874
XM_006717072.3
XP_006717135.1
26
3770
XM_006717074.3
XP_006717137.1
24
3364
XM_011518569.2
XP_011516871.1
27
3999
XM_011518570.2
XP_011516872.1
27
3888
XM_011518571.2
XP_011516873.1
27
4559
XM_011518572.2
XP_011516874.1
27
3738
XM_011518573.2
XP_011516875.1
27
3871
XM_011518574.2
XP_011516876.1
26
3798
XM_011518575.2
XP_011516877.1
26
3795
XM_011518576.2
XP_011516878.1
25
3540
XM_011518577.2
XP_011516879.1
25
3537
XM_011518578.2
XP_011516880.1
25
3474
XM_011518579.2
XP_011516881.1
24
3381
XM_017014633.1
XP_016870122.1
27
3884
XM_017014634.1
XP_016870123.1
27
3735
XM_017014635.1
XP_016870124.1
27
3789
XM_017014636.1
XP_016870125.1
27
3760
XM_017014637.1
XP_016870126.1
26
3732
XM_017014638.1
XP_016870127.1
25
3639
XM_017014639.1
XP_016870128.1
26
3633
GenBank mRNA
AK302763.1
-
16
1816
BC041710.1
-
24
3205
6051007
CR749354.1
-
24
3179
D21239.1
-
24
3243
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
blood vessel development
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of neuron projection development
single organismal cell-cell adhesion
Rap protein signal transduction
nerve growth factor signaling pathway
positive regulation of GTPase activity
negative regulation of Ras protein signal transduction
platelet-derived growth factor receptor signaling pathway
negative regulation of protein kinase B signaling
establishment of endothelial barrier
negative regulation of ERK1 and ERK2 cascade
cellular response to cAMP
negative regulation of canonical Wnt signaling pathway
regulation of cell junction assembly
cellular response to nerve growth factor stimulus
negative regulation of neural precursor cell proliferation
protein binding
Rap guanyl-nucleotide exchange factor activity
SH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0