Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306076.1 | NP_001293005.1 | 9 | 1230 | ||
NM_206886.4 | NP_996769.3 | 9 | 1230 | |||
XM_011541361.2 | XP_011539663.2 | 9 | 1439 | |||
XM_011541372.2 | XP_011539674.2 | 9 | 1439 | |||
XM_017001154.1 | XP_016856643.1 | 9 | 1439 | |||
XM_017001155.1 | XP_016856644.1 | 9 | 1439 | |||
XM_017001156.1 | XP_016856645.1 | 9 | 1409 | |||
XM_017001157.1 | XP_016856646.1 | 9 | 1439 | |||
XM_017001158.1 | XP_016856647.1 | 9 | 1439 | |||
XM_017001159.1 | XP_016856648.1 | 9 | 1439 | |||
XM_017001160.1 | XP_016856649.1 | 9 | 1439 | |||
XM_017001161.1 | XP_016856650.1 | 9 | 1439 | |||
XM_017001162.1 | XP_016856651.1 | 9 | 1152 | |||
XM_017001163.1 | XP_016856652.1 | 9 | 1706 | |||
XM_017001164.1 | XP_016856653.1 | 9 | 1250 | |||
XM_017001165.1 | XP_016856654.1 | 9 | 1439 | |||
XM_017001166.1 | XP_016856655.1 | 7 | 894 | |||
XM_017001167.1 | XP_016856656.1 | 7 | 829 | |||
XM_017001168.1 | XP_016856657.1 | 5 | 594 | |||
XM_017001169.1 | XP_016856658.1 | 9 | 1439 | |||
GenBank mRNA | AK126045.1 | - | 3 | 298 | ||
BX538286.1 | - | 2 | 221 |