Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004698.2 | NP_004689.1 | 2 | 290 | ||
XM_011510129.1 | XP_011508431.1 | 2 | 236 | |||
XM_011510131.2 | XP_011508433.1 | 2 | 230 | |||
XM_011510132.2 | XP_011508434.1 | 2 | 230 | |||
XM_017002791.1 | XP_016858280.1 | 2 | 230 | |||
GenBank mRNA | AF001947.1 | - | 1 | 186 | ||
AF016370.1 | - | 2 | 200 | |||
AK222825.1 | - | 2 | 216 | |||
AK297139.1 | - | 2 | 174 | |||
AK297265.1 | - | 2 | 216 | |||
AK299980.1 | - | 2 | 216 | |||
AK309330.1 | - | 2 | 222 | |||
AK311653.1 | - | 2 | 219 | |||
AK312891.1 | - | 2 | 211 | |||
BC000184.2 | - | 1 | 177 | 3351519 | ||
BC001954.1 | - | 1 | 185 | 2905308 | ||
DC407158.1 | - | 2 | 290 |