Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAB27A SIRNA Assays ›
          Gene Symbol:
          RAB27A
          Gene Name:
          RAB27A, member RAS oncogene family
          Gene Aliases:
          GS2, HsT18676, RAB27, RAM
          Chromosome Location:
          Chr.15: 55202966 - 55291188 on Build GRCh38
          UniGene ID:
          Hs.626665
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 14981

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAB27A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004580.4 NP_004571.2 3, 4 494
          NM_183234.2 NP_899057.1 4, 5 475
          NM_183235.2 NP_899058.1 4, 5 484
          NM_183236.2 NP_899059.1 4, 5 435
          XM_005254576.4 XP_005254633.1 3, 4 1737
          XM_011521852.1 XP_011520154.1 6, 7 698
          XM_011521854.1 XP_011520156.1 5, 6 571
          XM_011521855.2 XP_011520157.1 5, 6 1960
          XM_011521856.2 XP_011520158.1 3, 4 367
          GenBank mRNA AF125393.1 - 4, 5 475
          AF498953.1 - 2, 3 242
          AK312836.1 - 4, 5 482
          BC107680.1 - 4, 5 435 5416688
          BC132800.1 - 4, 5 420 40146822
          BC136423.1 - 4, 5 420 9020412
          BM014705.1 - 4, 5 435
          CR457076.1 - 2, 3 242
          CR536496.1 - 2, 3 242
          CR541693.1 - 2, 3 242
          DA182503.1 - 3, 4 494
          DA553342.1 - 4, 5 516
          U38654.3 - 3, 4 487
          U57094.1 - 4, 5 475

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001301230, NM_023635, NM_001301232, CV784419, AB046693, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting
          exocytosis
          small GTPase mediated signal transduction
          blood coagulation
          positive regulation of gene expression
          antigen processing and presentation
          melanocyte differentiation
          melanosome localization
          melanosome transport
          multivesicular body organization
          cytotoxic T cell degranulation
          natural killer cell degranulation
          positive regulation of exocytosis
          synaptic vesicle transport
          positive regulation of phagocytosis
          multivesicular body sorting pathway
          complement-dependent cytotoxicity
          positive regulation of regulated secretory pathway
          positive regulation of reactive oxygen species biosynthetic process
          positive regulation of constitutive secretory pathway
          exosomal secretion
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          protein domain specific binding
          myosin V binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0