Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZFP36L1 SIRNA Assays ›
          Gene Symbol:
          ZFP36L1
          Gene Name:
          ZFP36 ring finger protein like 1
          Gene Aliases:
          BRF1, Berg36, ERF-1, ERF1, RNF162B, TIS11B, cMG1
          Chromosome Location:
          Chr.14: 68787655 - 68796243 on Build GRCh38
          UniGene ID:
          Hs.707091
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 149889

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZFP36L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001244701.1 NP_001231630.1 1 53
          GenBank mRNA DA633985.1 - 1 53
          FY210721.1 - 1 24

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          vasculogenesis
          proepicardium development
          regulation of transcription, DNA-templated
          apoptotic process
          heart development
          cell proliferation
          response to wounding
          regulation of gene expression
          regulation of keratinocyte proliferation
          phosphatidylinositol 3-kinase signaling
          neural tube development
          nuclear-transcribed mRNA catabolic process, deadenylation-independent decay
          regulation of mRNA 3'-end processing
          cellular response to insulin stimulus
          T cell differentiation in thymus
          multicellular organism growth
          p38MAPK cascade
          regulation of mRNA stability
          protein kinase B signaling
          cellular response to fibroblast growth factor stimulus
          positive regulation of fat cell differentiation
          regulation of keratinocyte differentiation
          negative regulation of erythrocyte differentiation
          positive regulation of monocyte differentiation
          regulation of myoblast differentiation
          mesendoderm development
          mRNA transport
          chorio-allantoic fusion
          spongiotrophoblast layer development
          3'-UTR-mediated mRNA destabilization
          ERK1 and ERK2 cascade
          cellular response to cAMP
          cellular response to tumor necrosis factor
          cellular response to epidermal growth factor stimulus
          cellular response to peptide hormone stimulus
          cellular response to glucocorticoid stimulus
          cellular response to hypoxia
          cellular response to salt stress
          cellular response to transforming growth factor beta stimulus
          regulation of stem cell proliferation
          cellular response to raffinose
          positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          regulation of keratinocyte apoptotic process
          positive regulation of intracellular mRNA localization
          cellular response to phorbol 13-acetate 12-myristate
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          mRNA binding
          mRNA 3'-UTR binding
          protein binding
          mRNA 3'-UTR AU-rich region binding
          poly(A) RNA binding
          metal ion binding
          14-3-3 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0