siRNA Details
Gene Symbol: Rock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009071.2 | NP_033097.1 | 6, 7 | 1158 | ||
GenBank mRNA | AK149793.1 | - | 6, 7 | 1413 | ||
BC057154.1 | - | 6, 7 | 1158 | |||
BC066873.1 | - | 6, 7 | 835 | |||
U58512.1 | - | 6, 7 | 683 |

More Information
Other Species Targeted

Panther Classification:
