siRNA Details
Gene Symbol: Polr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009086.2 | NP_033112.2 | 15 | 2873 | ||
GenBank mRNA | AK083732.1 | - | 15 | 2856 | ||
AK143501.1 | - | 15 | 2810 | |||
AK144616.1 | - | 15 | 2860 | |||
AK156712.1 | - | 15 | 2863 | |||
BC060656.1 | - | 15 | 2803 | 30355130 | ||
U58280.1 | - | 15 | 2837 |

More Information
Other Species Targeted

Panther Classification:
Molecular Function -





