Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: St6galnac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009180.3 | NP_033206.2 | 9 | 1137 | ||
GenBank mRNA | AK004613.1 | - | 9 | 1138 | ||
AK079216.1 | - | 1295 | ||||
AK136381.1 | - | 1140 | ||||
BC003276.1 | - | 9 | 2039 | |||
BC010208.1 | - | 1089 | ||||
BC023076.1 | - | 3 | 1585 | |||
EU937514.1 | - | 3 | 1584 | |||
X93999.1 | - | 9 | 1101 |