Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009189.3 | NP_033215.2 | 2 | 863 | ||
GenBank mRNA | AK031122.1 | - | 2 | 618 | ||
AK131925.1 | - | 2 | 864 | |||
AK144753.1 | - | 2 | 874 | |||
BC023304.1 | - | 2 | 848 | 4188451 | ||
X80339.1 | - | 2 | 544 |