Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004615.3 | NP_004606.2 | 7, 8 | 828 | ||
GenBank mRNA | AK293178.1 | - | 8, 9 | 813 | ||
AK293534.1 | - | 9, 10 | 982 | |||
AK293552.1 | - | 8, 9 | 893 | |||
BC018036.1 | - | 7, 8 | 787 | 4801388 | ||
D10653.1 | - | 7, 8 | 768 | |||
D29808.1 | - | 7, 8 | 754 | |||
L10373.1 | - | 7, 8 | 812 |