Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BRD4 SIRNA Assays ›
          Gene Symbol:
          BRD4
          Gene Name:
          bromodomain containing 4
          Gene Aliases:
          CAP, HUNK1, HUNKI, MCAP
          Chromosome Location:
          Chr.19: 15236836 - 15332543 on Build GRCh38
          UniGene ID:
          Hs.187763
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1512
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BRD4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014299.2 NP_055114.1 3 599
          NM_058243.2 NP_490597.1 3 599
          XM_011527854.1 XP_011526156.1 3 664
          XM_011527855.1 XP_011526157.1 3 665
          XM_011527856.2 XP_011526158.1 3 664
          GenBank mRNA AF386649.1 - 3 599
          AK307664.1 - 3 489
          BC000156.1 - 3 445
          BC030158.2 - 3 466
          BC035266.1 - 3 670 4837491
          BC038988.1 - 3 640
          BC047888.1 - 3 653
          BC067129.1 - 3 621
          BC091649.1 - 3 637
          Y12059.1 - 3 599

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001100903, XM_006241074, XM_008765173, XM_006241073, XM_008765171, and others.
          This siRNA also targets the following transcripts in Mouse: NM_020508, NM_198094, NM_001286630, BC013666, BC082782, and others.

          Matching Taqman Gene Expression Assay:


          Hs00204762_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription involved in G1/S transition of mitotic cell cycle
          transcription, DNA-templated
          cellular response to DNA damage stimulus
          positive regulation of G2/M transition of mitotic cell cycle
          viral process
          covalent chromatin modification
          positive regulation of transcription elongation from RNA polymerase II promoter
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of transcription from RNA polymerase II promoter
          regulation of inflammatory response
          regulation of phosphorylation of RNA polymerase II C-terminal domain
          p53 binding
          chromatin binding
          protein binding
          lysine-acetylated histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline