Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011161.5 | NP_035291.4 | 12 | 1705 | ||
GenBank mRNA | AK144846.1 | - | 12 | 1678 | ||
AK161190.1 | - | 10 | 1880 | |||
AK161562.1 | - | 12 | 1657 | |||
BC057211.1 | - | 13 | 1347 | |||
BC064737.1 | - | 12 | 1654 | 30112690 | ||
BC092526.1 | - | 12 | 1705 | 6593321 |