Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202445.1 | NP_001189374.1 | 12 | 2996 | ||
NM_001202446.1 | NP_001189375.1 | 12 | 2921 | |||
NM_011162.5 | NP_035292.2 | 12 | 2704 | |||
GenBank mRNA | AF003115.1 | - | 12 | 2563 | ||
AF054611.1 | - | 12 | 2704 | |||
AK147033.1 | - | 12 | 2923 | |||
BC072578.1 | - | 12 | 2888 | 6402485 |