Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GM21704 SIRNA Assays ›
          Gene Symbol:
          Gm21704 Gm10256 Gm10352 Gm21677 Gm21693 Gm21708 Gm3376 Gm4064 Rbmy
          Gene Name:
          predicted gene, 21704
          predicted gene 10256
          predicted gene 10352
          predicted gene, 21677
          predicted gene, 21693
          predicted gene, 21708
          predicted gene 3376
          predicted gene 4064
          RNA binding motif protein, Y chromosome
          Gene Aliases:
          -
          ENSMUSG00000069031
          ENSMUSG00000071960
          -
          -
          -
          Rbmy1b
          -
          RBM, Rbm1, Rbm1-rs1, Rbmy1a1, Rbmy1a1-rs1, Rbmy1b
          Chromosome Location:
          Chr.Y: 3367143 - 3378783 on Build GRCm38
          Chromosome Location:
          Chr.Y: 2862139 - 2873496 on Build GRCm38
          Chromosome Location:
          Chr.Y: 2900989 - 2912210 on Build GRCm38
          Chromosome Location:
          Chr.Y: 3294972 - 3306449 on Build GRCm38
          Chromosome Location:
          Chr.Y: 3326746 - 3345434 on Build GRCm38
          Chromosome Location:
          Chr.Y: 3399184 - 3410167 on Build GRCm38
          Chromosome Location:
          Chr.Y: 3771673 - 3783271 on Build GRCm38
          Chromosome Location:
          Chr.Y: 2785165 - 2796224 on Build GRCm38
          Chromosome Location:
          Chr.Y: 2830680 - 2841858 on Build GRCm38
          UniGene ID:
          Mm.4711
          Mm.4711
          Mm.4711
          Mm.4711
          Mm.4711
          Mm.4711
          Mm.491824
          Mm.414207
          Mm.4711
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 151434

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gm21704
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270515.1 NP_001257444.1 1013
          Gene Symbol: Gm10256
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270510.1 NP_001257439.1 1013
          Gene Symbol: Gm10352
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270511.1 NP_001257440.1 1013
          Gene Symbol: Gm21677
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270513.1 NP_001257442.1 1013
          Gene Symbol: Gm21693
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270514.1 NP_001257443.1 1013
          Gene Symbol: Gm21708
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270516.1 NP_001257445.1 9 1013
          Gene Symbol: Gm3376
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270512.1 NP_001257441.1 9 1013
          Gene Symbol: Gm4064
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270518.1 NP_001257447.1 9 1013
          Gene Symbol: Rbmy
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166384.1 NP_001159856.1 9 1013
          NM_011253.2 NP_035383.2 10 1164
          GenBank mRNA AK016258.1 - 1015
          BC150670.1 - 994 9087581
          U36929.1 - 1005
          Y15131.1 - 10 1153

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          hematopoietic progenitor cell differentiation
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0