Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RELN SIRNA Assays ›
Gene Symbol:
Reln
Gene Name:
reelin
Gene Aliases:
reeler, rl
Chromosome Location:
Chr.5: 21884454 - 22344705 on Build GRCm38
UniGene ID:
Mm.425236
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
151444
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Reln
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_011261.2
NP_035391.2
10, 11
1440
GenBank mRNA
AK082447.1
-
10, 11
1441
U24703.1
-
10, 11
1425
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix glycoprotein
extracellular matrix protein
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis involved in differentiation
neuron migration
proteolysis
cell adhesion
multicellular organism development
axon guidance
central nervous system development
brain development
long-term memory
locomotory behavior
associative learning
glial cell differentiation
regulation of gene expression
positive regulation of neuron projection development
positive regulation of phosphatidylinositol 3-kinase signaling
dendrite development
cell migration
peptidyl-tyrosine phosphorylation
spinal cord patterning
dentate gyrus development
hippocampus development
cerebral cortex tangential migration
layer formation in cerebral cortex
cerebral cortex development
forebrain development
positive regulation of TOR signaling
positive regulation of CREB transcription factor activity
protein localization to synapse
reelin-mediated signaling pathway
thyroid hormone metabolic process
positive regulation of protein kinase activity
response to pain
positive regulation of peptidyl-tyrosine phosphorylation
regulation of behavior
modulation of synaptic transmission
positive regulation of small GTPase mediated signal transduction
positive regulation of synaptic transmission, glutamatergic
positive regulation of dendritic spine morphogenesis
positive regulation of protein tyrosine kinase activity
positive regulation of synapse maturation
postsynaptic density assembly
NMDA glutamate receptor clustering
postsynaptic density protein 95 clustering
receptor localization to synapse
lateral motor column neuron migration
positive regulation of long-term synaptic potentiation
positive regulation of lateral motor column neuron migration
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of excitatory postsynaptic potential
positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
protein binding
peptidase activity
serine-type peptidase activity
hydrolase activity
metal ion binding
lipoprotein particle receptor binding
very-low-density lipoprotein particle receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0