siRNA Details
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113559.2 | NP_001107031.2 | 7 | 1033 | ||
NM_001243163.1 | NP_001230092.1 | 7 | 925 | |||
NM_011444.3 | NP_035574.2 | 7 | 1030 | |||
GenBank mRNA | AB006330.1 | - | 7 | 942 | ||
AJ010604.1 | - | 7 | 915 | |||
AK029047.1 | - | 10 | 1175 | |||
AK081802.1 | - | 7 | 1322 | |||
BC110478.2 | - | 7 | 917 | 40047865 | ||
KF793916.1 | - | 7 | 731 |

More Information
Other Species Targeted

Panther Classification:
