Search
Search

Rpn2| Gene Symbol: Rpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019642.4 | NP_062616.2 | 3 | 444 | ||
| GenBank mRNA | AK004968.1 | - | 3 | 445 | ||
| AK018761.2 | - | 3 | 483 | |||
| AK153952.1 | - | 3 | 305 | |||
| AK161624.1 | - | 3 | 255 | |||
| AK166778.1 | - | 3 | 230 | |||
| AK167864.1 | - | 3 | 226 | |||
| AK167866.1 | - | 3 | 226 | |||
| AK168221.1 | - | 3 | 226 | |||
| BC010509.1 | - | 2 | 217 | |||
| BC046806.1 | - | 3 | 472 | 6829209 | ||
| D31717.1 | - | 2 | 230 | |||