Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009503.4 | NP_033529.3 | 2 | 404 | ||
GenBank mRNA | AK028264.1 | - | 2 | 333 | ||
AK030751.1 | - | 2 | 235 | |||
AK146109.1 | - | 2 | 301 | |||
AK149931.1 | - | 2 | 297 | |||
AK151109.1 | - | 2 | 298 | |||
AK151418.1 | - | 2 | 298 | |||
AK153012.1 | - | 2 | 334 | |||
AK153249.1 | - | 2 | 375 | |||
AK159177.1 | - | 2 | 333 | |||
AK159475.1 | - | 2 | 332 | |||
AK159509.1 | - | 2 | 376 | |||
AK160889.1 | - | 2 | 221 | |||
AK167794.1 | - | 2 | 352 | |||
AK169140.1 | - | 2 | 306 | |||
AK190461.1 | - | 2 | 92 | |||
AK194156.1 | - | 2 | 92 | |||
AK194828.1 | - | 2 | 92 | |||
AK194993.1 | - | 2 | 92 | |||
AK197087.1 | - | 2 | 92 | |||
AK197232.1 | - | 2 | 92 | |||
AK199977.1 | - | 2 | 92 | |||
AK202627.1 | - | 2 | 92 | |||
AK202683.1 | - | 2 | 92 | |||
AK203987.1 | - | 2 | 92 | |||
AK205011.1 | - | 2 | 92 | |||
AK205885.1 | - | 2 | 92 | |||
AK211805.1 | - | 2 | 92 | |||
AK212799.1 | - | 2 | 92 | |||
AK215162.1 | - | 2 | 92 | |||
AK218422.1 | - | 2 | 92 | |||
BC043053.1 | - | 2 | 308 | 6418662 | ||
BC049114.1 | - | 2 | 352 | 6810751 | ||
BY732145.1 | - | 2 | 404 | |||
Z14044.1 | - | 2 | 236 |