Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290299.1 | NP_001277228.1 | 10 | 1935 | ||
NM_144935.1 | NP_659184.1 | 9 | 1891 | |||
GenBank mRNA | AK082453.1 | - | 10 | 1898 | ||
AY438023.1 | - | 9 | 1891 | |||
BC018242.1 | - | 9 | 1891 | 4503683 | ||
BC023082.1 | - | 10 | 1912 | 5357160 |