Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145711.4 | NP_663757.3 | 5 | 1080 | ||
GenBank mRNA | AF472514.1 | - | 5 | 876 | ||
AK029694.1 | - | 5 | 877 | |||
AK038671.1 | - | 5 | 1083 | |||
AK133397.1 | - | 5 | 877 | |||
AK161578.1 | - | 5 | 866 | |||
BC038873.1 | - | 1 | 96 | |||
BC080732.1 | - | 5 | 997 | 6518356 |