Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSPOAP1 SIRNA Assays ›
Gene Symbol:
TSPOAP1
Gene Name:
TSPO associated protein 1
Gene Aliases:
BZRAP1, PBR-IP, PRAX-1, PRAX1, RIM-BP1, RIMBP1
Chromosome Location:
Chr.17: 58301227 - 58329030 on Build GRCh38
UniGene ID:
Hs.112499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15280
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSPOAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261835.1
NP_001248764.1
1
1053
NM_004758.3
NP_004749.2
1
1053
NM_024418.2
NP_077729.1
1
1053
XM_005257807.2
XP_005257864.1
1
1057
XM_006722173.2
XP_006722236.1
1
1057
XM_006722174.2
XP_006722237.1
1
1057
XM_006722175.2
XP_006722238.1
1
1057
XM_006722176.2
XP_006722239.1
1
1057
XM_017025326.1
XP_016880815.1
1
1057
XM_017025327.1
XP_016880816.1
2
205
XM_017025328.1
XP_016880817.1
1
1057
XM_017025329.1
XP_016880818.1
1
1057
XM_017025330.1
XP_016880819.1
1
1057
XM_017025331.1
XP_016880820.1
1
1057
XM_017025332.1
XP_016880821.1
1
1057
XM_017025333.1
XP_016880822.1
1
1057
XM_017025334.1
XP_016880823.1
1
1057
GenBank mRNA
AB014512.2
-
1
1022
AF039571.1
-
1
379
BC146852.1
-
1
346
9056780
BC150279.1
-
1
1022
9007261
BC171787.1
-
1
346
9054441
KJ534781.1
-
1
182
KJ534782.1
-
1
182
KJ534783.1
-
1
182
KJ534784.1
-
1
182
KJ534785.1
-
1
182
KJ534786.1
-
1
182
KJ534787.1
-
1
182
KJ534788.1
-
1
182
KJ534789.1
-
1
182
KJ534790.1
-
1
182
KJ535016.1
-
1
182
KJ535062.1
-
1
182
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neurotransmitter secretion
biological_process
glutamate secretion
protein binding
benzodiazepine receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0