Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PICALM SIRNA Assays ›
          Gene Symbol:
          Picalm
          Gene Name:
          phosphatidylinositol binding clathrin assembly protein
          Gene Aliases:
          CALM, CLTH, PAP180, fit-1, fit1, mKIAA4114
          Chromosome Location:
          Chr.7: 90130220 - 90209447 on Build GRCm38
          UniGene ID:
          Mm.235175
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 152804

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Picalm
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252520.1 NP_001239449.1 8 1133
          NM_001252521.1 NP_001239450.1 8 1133
          NM_001252522.1 NP_001239451.1 8 1133
          NM_001252523.1 NP_001239452.1 8 1133
          NM_001252524.1 NP_001239453.1 8 1133
          NM_146194.4 NP_666306.2 8 1133
          GenBank mRNA AK085472.2 - 5 367
          AK144960.1 - 8 872
          AK162360.1 - 8 826
          AK165066.1 - 3 244
          AK170373.1 - 8 1153
          AK220291.1 - 8 849
          AY206701.1 - 8 817
          BC011470.1 - 8 843 2651109
          BC021491.1 - 8 1082 5345558
          BC023843.1 - 8 881 5346041
          BC025566.1 - 8 1156 5322606
          BC027116.1 - 5 444
          BC057683.1 - 8 1084 3671962

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053554, XM_006229676, XM_006229674, XM_006229681, XM_006229679, and others.

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          receptor-mediated endocytosis
          multicellular organism development
          axonogenesis
          cell proliferation
          negative regulation of gene expression
          synaptic vesicle maturation
          vesicle-mediated transport
          endosomal transport
          hemopoiesis
          regulation of endocytosis
          receptor internalization
          regulation of protein localization
          cargo loading into vesicle
          positive regulation of transcription, DNA-templated
          negative regulation of receptor-mediated endocytosis
          clathrin coat assembly
          dendrite morphogenesis
          regulation of protein transport
          iron ion homeostasis
          clathrin-mediated endocytosis
          iron ion import into cell
          positive regulation of neuron death
          positive regulation of beta-amyloid formation
          regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          positive regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
          phospholipid binding
          1-phosphatidylinositol binding
          SH3 domain binding
          clathrin binding
          clathrin heavy chain binding
          clathrin adaptor activity
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0