Search
Search

Rfwd3| Gene Symbol: Rfwd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146218.4 | NP_666330.2 | 2 | 135 | ||
| GenBank mRNA | AK137499.1 | - | 2 | 115 | ||
| AK137698.1 | - | 2 | 98 | |||
| AK139032.1 | - | 2 | 115 | |||
| AK146451.1 | - | 2 | 105 | |||
| BC023694.1 | - | 2 | 140 | 5344528 | ||
| BC096602.1 | - | 2 | 98 | 4954068 | ||