Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291448.1 | NP_001278377.1 | 2, 3 | 466 | ||
NM_172291.2 | NP_758495.1 | 2, 3 | 466 | |||
GenBank mRNA | AK054231.1 | - | 2, 3 | 358 | ||
AK144759.1 | - | 2, 3 | 392 | |||
AK163722.1 | - | 2, 3 | 325 | |||
BC024806.1 | - | 2, 3 | 336 | 5361053 |