Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172806.2 | NP_766394.2 | 3, 4 | 527 | ||
GenBank mRNA | AK033160.1 | - | 2, 3 | 440 | ||
AK122522.1 | - | 1, 2 | 177 | |||
AK134271.1 | - | 2, 3 | 378 | |||
BC041669.1 | - | 3, 4 | 527 | 3482657 | ||
BC057303.1 | - | 2, 3 | 246 | 5702276 |