Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172876.2 | NP_766464.1 | 2 | 660 | ||
GenBank mRNA | AK053051.1 | - | 2 | 661 | ||
BC096468.1 | - | 2 | 646 | 3665952 | ||
BC121828.1 | - | 2 | 661 | 40107316 | ||
BC121829.1 | - | 2 | 638 | 40107315 |