Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172896.2 | NP_766484.1 | 9 | 2906 | ||
GenBank mRNA | AK054378.1 | - | 9 | 2797 | ||
AK087843.1 | - | 9 | 2907 | |||
AK135745.1 | - | 6 | 1720 | |||
AK136205.1 | - | 10 | 2882 | |||
AY355341.1 | - | 9 | 2907 | |||
AY596200.1 | - | 7 | 2603 | |||
BC141342.1 | - | 9 | 2806 | 9055954 | ||
BC141343.1 | - | 9 | 2806 | 9055955 |