Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173026.2 | NP_766614.2 | 5 | 1957 | ||
GenBank mRNA | AK033794.1 | - | 2 | 638 | ||
AK048360.1 | - | 2 | 2238 |