Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SRSF11 SIRNA Assays ›
Gene Symbol:
SRSF11
Gene Name:
serine and arginine rich splicing factor 11
Gene Aliases:
NET2, SFRS11, dJ677H15.2, p54
Chromosome Location:
Chr.1: 70205682 - 70252020 on Build GRCh38
UniGene ID:
Hs.479693
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15376
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SRSF11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190987.1
NP_001177916.1
13
2168
NM_004768.3
NP_004759.1
13
2174
XM_005271338.3
XP_005271395.1
13
2186
XM_005271339.3
XP_005271396.1
13
2188
XM_011542427.2
XP_011540729.1
12
2327
XM_011542428.2
XP_011540730.1
12
5584
XM_011542431.2
XP_011540733.1
13
3348
XM_017002811.1
XP_016858300.1
13
2183
XM_017002812.1
XP_016858301.1
13
2165
XM_017002813.1
XP_016858302.1
13
2164
XM_017002814.1
XP_016858303.1
11
3317
XM_017002815.1
XP_016858304.1
13
4929
XM_017002816.1
XP_016858305.1
14
5063
XM_017002817.1
XP_016858306.1
14
4637
XM_017002818.1
XP_016858307.1
13
2465
XM_017002819.1
XP_016858308.1
14
2574
XM_017002820.1
XP_016858309.1
13
5289
XM_017002821.1
XP_016858310.1
13
2462
XM_017002822.1
XP_016858311.1
11
3659
XM_017002823.1
XP_016858312.1
12
3004
XM_017002824.1
XP_016858313.1
14
5042
XM_017002825.1
XP_016858314.1
14
4616
XM_017002826.1
XP_016858315.1
13
2444
XM_017002827.1
XP_016858316.1
14
2553
XM_017002828.1
XP_016858317.1
15
4387
XM_017002829.1
XP_016858318.1
15
4423
XM_017002830.1
XP_016858319.1
13
5545
XM_017002831.1
XP_016858320.1
14
4613
XM_017002832.1
XP_016858321.1
13
2441
XM_017002833.1
XP_016858322.1
14
2550
XM_017002834.1
XP_016858323.1
12
3283
GenBank mRNA
AB062486.1
-
1
486
AL832279.1
-
10
2433
BC040436.1
-
13
2126
5518580
BX537679.1
-
2
2904
BX640645.1
-
11
3411
BX641004.1
-
9
1594
M74002.1
-
13
2133
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA processing
RNA export from nucleus
mRNA export from nucleus
RNA splicing
mRNA 3'-end processing
nucleotide binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0