Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177721.4 | NP_808389.2 | 1 | 2463 | ||
GenBank mRNA | AK083089.1 | - | 1 | 443 | ||
AK161331.1 | - | 1 | 165 | |||
BC026487.1 | - | 1 | 365 | |||
BC056759.1 | - | 1 | 2442 | 6518485 | ||
BC139395.1 | - | 1 | 2463 | 8862417 | ||
BC139396.1 | - | 1 | 2463 | 8862418 |